ENA321762221 Enamine

oxan-3-one

  • MDL number MFCD00182426
  • Cas Number : 23462-75-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 70.4
Storage temp. -20
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-72286

Safety Information

signalword Warning
pcodes P210,P233,P240,P241,P242,P243,P280,P303+P361+P353,P370+P378,P403+P235,P501
MSDS https://www.enaminestore.com/catalog/EN300-72286
pictograms GHS02
hstatements H226
Availability details for
Quantity
  • Availability
  • Add to Cart