ENA321762223 Enamine

2-chloro-N-(3-oxo-3,4-dihydro-2H-1,4-benzoxazin-6-yl)acetamide

  • MDL number MFCD10061339
  • Cas Number : 496056-85-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-72319

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72319
Availability details for
Quantity
  • Availability
  • Add to Cart