ENA321762224 Enamine

imidazo[1,2-a]pyrazin-8-amine

  • MDL number MFCD09835034
  • Cas Number : 117718-88-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-72346

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72346
Availability details for
Quantity
  • Availability
  • Add to Cart