ENA321762229 Enamine

(1S)-1-cyclopropylethan-1-ol

  • MDL number MFCD08702991
  • Cas Number : 55637-37-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72586

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-72586
pictograms GHS02
Availability details for
Quantity
  • Availability
  • Add to Cart