ENA321762233 Enamine

7-(1,3-dioxaindan-5-yl)-1-methyl-2,4-dioxo-1H,2H,3H,4H-pyrido[2,3-d]pyrimidine-5-carboxylic acid

  • MDL number MFCD08258477
  • Cas Number : 879303-83-0
  • Molecular Weight : 341.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-72671

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72671
Availability details for
Quantity
  • Availability
  • Add to Cart