ENA321762242 Enamine

2-[(morpholine-4-sulfonyl)amino]ethan-1-ol

  • MDL number MFCD12185471
  • Cas Number : 1156254-42-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72798

Safety Information

msds https://www.enaminestore.com/catalog/EN300-72798
Availability details for
Quantity
  • Availability
  • Add to Cart