ENA321762248 Enamine

2-chloro-N-methyl-N-(2-methylcyclohexyl)acetamide

  • MDL number MFCD12798885
  • Cas Number : 1183795-70-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-46611

Safety Information

msds https://www.enaminestore.com/catalog/EN300-46611
Availability details for
Quantity
  • Availability
  • Add to Cart