ENA321762249 Enamine

2-chloro-1-(4-methyl-decahydroquinolin-1-yl)ethan-1-one

  • MDL number MFCD12762657
  • Cas Number : 76395-69-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-46659

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-46659
Availability details for
Quantity
  • Availability
  • Add to Cart