ENA321762250 Enamine

2-chloro-N-methyl-N-(2,2,2-trifluoroethyl)acetamide

  • MDL number MFCD12798886
  • Cas Number : 1179829-40-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-46669

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-46669
Availability details for
Quantity
  • Availability
  • Add to Cart