ENA321762253 Enamine

5-chloro-1,3-dimethyl-1H-pyrazole-4-carbonitrile

  • MDL number MFCD01569082
  • Cas Number : 96286-02-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73017

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73017
Availability details for
Quantity
  • Availability
  • Add to Cart