ENA321762266 Enamine

2-chloro-N-(4,5,6,7-tetrahydro-1,3-benzothiazol-2-yl)acetamide

  • MDL number MFCD00719690
  • Cas Number : 22455-51-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-46958

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-46958
Availability details for
Quantity
  • Availability
  • Add to Cart