ENA321762267 Enamine

1-(piperidin-2-yl)propan-1-ol

  • MDL number MFCD18785452
  • Cas Number : 63401-12-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 89-90
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-73193

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73193
Availability details for
Quantity
  • Availability
  • Add to Cart