ENA321762269 Enamine

1-[1-(3-chlorophenyl)-5-methyl-1H-pyrazol-4-yl]ethan-1-one

  • MDL number MFCD00112456
  • Cas Number : 10021-79-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73216

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73216
Availability details for
Quantity
  • Availability
  • Add to Cart