ENA321762271 Enamine

2-[(3-formylphenoxy)methyl]benzoic acid

  • MDL number MFCD12469724
  • Cas Number : 1154270-79-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-73275

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73275
Availability details for
Quantity
  • Availability
  • Add to Cart