ENA321762278 Enamine

2-chloro-N-{4H,5H,6H-cyclopenta[d][1,3]thiazol-2-yl}acetamide

  • MDL number MFCD14649122
  • Cas Number : 1249003-51-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-47102

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-47102
Availability details for
Quantity
  • Availability
  • Add to Cart