ENA321762279 Enamine

6-(bromomethyl)imidazo[2,1-b][1,3]thiazole hydrobromide

  • MDL number MFCD00725035
  • Cas Number : 330196-49-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-01830

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-01830
Availability details for
Quantity
  • Availability
  • Add to Cart