ENA321762280 Enamine

2-[(2-methylpropoxy)methyl]oxirane

  • MDL number MFCD00055638
  • Cas Number : 3814-55-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Flash Point 49
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-22419

Safety Information

signalword Warning
safety_statement 26-37/39
msds https://www.enaminestore.com/catalog/EN300-22419
pictograms GHS02,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart