ENA321762284 Enamine

1-ethyl-1,2,3,4-tetrahydroquinoxaline

  • MDL number MFCD01684956
  • Cas Number : 73855-46-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-73359

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73359
Availability details for
Quantity
  • Availability
  • Add to Cart