ENA321762290 Enamine

cyclopentyl(4-fluorophenyl)methanone

  • MDL number MFCD03841310
  • Cas Number : 31545-25-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73387

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73387
Availability details for
Quantity
  • Availability
  • Add to Cart