ENA321762293 Enamine

1-(4-methylbenzenesulfonyl)-1H-pyrrole-3-carboxylic acid

  • MDL number MFCD00145013
  • Cas Number : 106058-86-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 202-206
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-73409

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73409
Availability details for
Quantity
  • Availability
  • Add to Cart