ENA321762299 Enamine

6-methyl-2-oxo-1,2,5,6,7,8-hexahydroquinoline-3-carbonitrile

  • MDL number MFCD16070467
  • Cas Number : 1247158-33-3
  • Molecular Weight : 188.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-73482

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73482
Availability details for
Quantity
  • Availability
  • Add to Cart