ENA321762306 Enamine

2-chloro-N-[2-oxo-2-(thiophen-2-yl)ethyl]acetamide

  • MDL number MFCD11557976
  • Cas Number : 70996-66-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-73511

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73511
Availability details for
Quantity
  • Availability
  • Add to Cart