ENA321762308 Enamine

4-[4-(cyanomethyl)phenoxy]benzoic acid

  • MDL number MFCD12452313
  • Cas Number : 1021144-57-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-73517

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73517
Availability details for
Quantity
  • Availability
  • Add to Cart