ENA321762312 Enamine

3-(4-aminophenoxy)benzamide

  • MDL number MFCD11133180
  • Cas Number : 284462-85-7
  • Molecular Weight : 228.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-73537

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73537
Availability details for
Quantity
  • Availability
  • Add to Cart