ENA321762320 Enamine

2-chloro-N-(2-cyclohexylethyl)acetamide

  • MDL number MFCD12798903
  • Cas Number : 1179762-09-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-47460

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-47460
Availability details for
Quantity
  • Availability
  • Add to Cart