ENA321762321 Enamine

4-phenyl-1H-imidazole-2-carbaldehyde

  • MDL number MFCD10696709
  • Cas Number : 56248-10-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73581

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73581
Availability details for
Quantity
  • Availability
  • Add to Cart