ENA321762322 Enamine

N-(2-bromoethyl)ethane-1-sulfonamide

  • MDL number MFCD11101724
  • Cas Number : 63132-88-7
  • Molecular Weight : 216.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-73582

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73582
Availability details for
Quantity
  • Availability
  • Add to Cart