ENA321762323 Enamine

1-(2,3-dihydro-1,4-benzodioxine-2-carbonyl)piperidin-4-amine hydrochloride

  • MDL number MFCD18483190
  • Cas Number : 1281025-81-7
  • Molecular Weight : 298.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73583

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73583
Availability details for
Quantity
  • Availability
  • Add to Cart