ENA321762324 Enamine

6-bromo-4-chloro-8-fluoro-2-methylquinoline

  • MDL number MFCD12115097
  • Cas Number : 1156277-76-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-73589

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73589
Availability details for
Quantity
  • Availability
  • Add to Cart