ENA321762326 Enamine

1-cyclohexylpiperidin-3-amine

  • MDL number MFCD14617572
  • Cas Number : 933713-15-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73603

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73603
Availability details for
Quantity
  • Availability
  • Add to Cart