ENA321762330 Enamine

2-chloro-1-{4-ethyl-4H,5H,6H,7H-thieno[3,2-c]pyridin-5-yl}ethan-1-one

  • MDL number MFCD12798430
  • Cas Number : 1182908-09-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-24633

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24633
Availability details for
Quantity
  • Availability
  • Add to Cart