ENA321762341 Enamine

3-(propan-2-yl)-2,3,4,5-tetrahydro-1H-1,4-benzodiazepine-2,5-dione

  • MDL number MFCD08741813
  • Cas Number : 24919-51-5
  • Molecular Weight : 218.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-73651

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73651
Availability details for
Quantity
  • Availability
  • Add to Cart