ENA321762355 Enamine

5-(thiophen-3-ylmethyl)-1,3,4-oxadiazol-2-amine

  • MDL number MFCD11170355
  • Cas Number : 1042654-57-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 151...153
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73680

Safety Information

msds https://www.enaminestore.com/catalog/EN300-73680
Availability details for
Quantity
  • Availability
  • Add to Cart