ENA321762360 Enamine

2-(2-chloroacetamido)-N,N-dimethylpropanamide

  • MDL number MFCD12142225
  • Cas Number : 1218689-32-7
  • Molecular Weight : 192.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-47657

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-47657
Availability details for
Quantity
  • Availability
  • Add to Cart