ENA321762367 Enamine

3-[(1H-1,3-benzodiazol-1-yl)methyl]aniline

  • MDL number MFCD09812505
  • Cas Number : 562803-83-2
  • Molecular Weight : 223.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-73700

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73700
Availability details for
Quantity
  • Availability
  • Add to Cart