ENA321762372 Enamine

4-[(4-methyl-1H-pyrazol-1-yl)methyl]aniline

  • MDL number MFCD12763842
  • Cas Number : 1183309-99-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-73709

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73709
Availability details for
Quantity
  • Availability
  • Add to Cart