ENA321762379 Enamine

7-hydroxy-3,4-dihydro-2H-1-benzopyran-3-carboxylic acid

  • MDL number MFCD08442197
  • Cas Number : 927802-07-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27493

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27493
Availability details for
Quantity
  • Availability
  • Add to Cart