ENA321762381 Enamine

2-(ethoxymethyl)oxirane

  • MDL number MFCD00085264

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Flash Point 47
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73725

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-73725
pictograms GHS02,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart