ENA321762385 Enamine

2-{[(benzyloxy)carbonyl]amino}-5-hydroxybenzoic acid

  • MDL number MFCD17170544
  • Cas Number : 214957-88-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 176...178
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73731

Safety Information

msds https://www.enaminestore.com/catalog/EN300-73731
Availability details for
Quantity
  • Availability
  • Add to Cart