ENA321762390 Enamine

4-cyclohexylbutan-2-ol

  • MDL number MFCD16091521
  • Cas Number : 10528-67-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73736

Safety Information

msds https://www.enaminestore.com/catalog/EN300-73736
Availability details for
Quantity
  • Availability
  • Add to Cart