ENA321762391 Enamine

2-(2,2,2-trifluoroethoxy)ethan-1-ol

  • MDL number MFCD00462508
  • Cas Number : 2358-54-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Flash Point 70
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73737

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-73737
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart