ENA321762393 Enamine

3-[(methylsulfanyl)methyl]aniline

  • MDL number MFCD11868819
  • Cas Number : 90607-33-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73739

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73739
Availability details for
Quantity
  • Availability
  • Add to Cart