ENA321762398 Enamine

1-benzyl-1H,6H,7H-pyrrolo[2,3-c]pyridin-7-one

  • MDL number MFCD18483204
  • Cas Number : 60290-20-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73745

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73745
Availability details for
Quantity
  • Availability
  • Add to Cart