ENA321762399 Enamine

5-methyl-1-phenyl-1H-1,2,4-triazole-3-carboxylic acid

  • MDL number MFCD11053232
  • Cas Number : 1016-57-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-73746

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73746
Availability details for
Quantity
  • Availability
  • Add to Cart