ENA321762408 Enamine

1-[(4-bromophenyl)methyl]pyrrolidine

  • MDL number MFCD06080622
  • Cas Number : 4897-55-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73758

Safety Information

msds https://www.enaminestore.com/catalog/EN300-73758
Availability details for
Quantity
  • Availability
  • Add to Cart