ENA321762409 Enamine

[1-(4-aminobutyl)pyrrolidin-2-yl]methanol

  • MDL number MFCD14660596
  • Cas Number : 1249116-36-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73760

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73760
Availability details for
Quantity
  • Availability
  • Add to Cart