ENA321762411 Enamine

6-(3-methoxybenzyl)-4-methyl-1,6-dihydropyrazolo[3,4-c]pyrazol-3-amine

  • MDL number MFCD09041065
  • Cas Number : 1280804-28-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73769

Safety Information

msds https://www.enaminestore.com/catalog/EN300-73769
Availability details for
Quantity
  • Availability
  • Add to Cart