ENA321762418 Enamine

4-(trifluoromethoxy)benzene-1-sulfonohydrazide

  • MDL number MFCD04123712
  • Cas Number : 685542-10-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-73781

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73781
Availability details for
Quantity
  • Availability
  • Add to Cart