ENA321762420 Enamine

[3-(3-methylbutoxy)phenyl]boronic acid

  • MDL number MFCD11183190
  • Cas Number : 1236189-95-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-73783

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73783
Availability details for
Quantity
  • Availability
  • Add to Cart