ENA321762421 Enamine

2-(4-aminopiperidin-1-yl)-N,N-dimethylacetamide

  • MDL number MFCD09046658
  • Cas Number : 882562-49-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73784

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73784
Availability details for
Quantity
  • Availability
  • Add to Cart